4-(2-Aminoethoxy)benzonitril 97%

Lieferant: Apollo Scientific

OR965062-1G OR965062-250MG OR965062-5G
APOSOR965062-1GEA 226 EUR
APOSOR965062-1G APOSOR965062-250MG APOSOR965062-5G
4-(2-Aminoethoxy)benzonitril 97%
4-(2-Aminoethoxy)benzonitril
Formel: C₉H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD16495887
CAS-Nummer: 67333-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR