7-Bromo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-2,3-dihydro-[1,4]dioxino[2,3-b]pyridine

OR965152-100MG OR965152-1G OR965152-250MG
APOSOR965152-100MGEA 177 EUR
APOSOR965152-100MG APOSOR965152-1G APOSOR965152-250MG
7-Bromo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine 97%
7-Bromo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
Formel: C₇H₆BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD15530247
CAS-Nummer: 95897-49-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR