2-(2-Methyl-1H-indol-1-yl)acetic acid 97%

Lieferant: Apollo Scientific

OR965293-10G OR965293-1G OR965293-5G
APOSOR965293-10GEA 456 EUR
APOSOR965293-10G APOSOR965293-1G APOSOR965293-5G
2-(2-Methyl-1H-indol-1-yl)acetic acid 97%
2-(2-Methyl-1H-indol-1-yl)acetic acid
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 86704-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR