2-Methyltetrahydrofuran-2-carboxylic acid 96%

Lieferant: Apollo Scientific

OR965333-1G OR965333-250MG OR965333-5G
APOSOR965333-1GEA 284 EUR
APOSOR965333-1G APOSOR965333-250MG APOSOR965333-5G
2-Methyltetrahydrofuran-2-carboxylic acid 96%
2-Methyltetrahydrofuran-2-carboxylic acid
Formel: C₆H₁₀O₃
Molecular Weight: 130.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06809613
CAS-Nummer: 61449-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR