3-tert-Butylsulphanyl-pyridin-2-ylamine 95%
Lieferant: Apollo Scientific
OR965345-1G
OR965345-250MG
APOSOR965345-1GEA
271
EUR
APOSOR965345-1G
APOSOR965345-250MG
3-tert-Butylsulphanyl-pyridin-2-ylamine 95%
3-tert-Butylsulphanyl-pyridin-2-ylamine
Formel:
C₉H₁₄N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663516 CAS-Nummer: 551950-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...