Ethyl 2-(pyrimidin-2-yl)acetate 98%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-(pyrimidin-2-yl)acetate, 2-Pyrimidineacetic acid ethyl ester
OR965521-10G
OR965521-1G
OR965521-5G
APOSOR965521-10GEA
337
EUR
APOSOR965521-10G
APOSOR965521-1G
APOSOR965521-5G
Ethyl 2-(pyrimidin-2-yl)acetate 98%
Ethyl-2-(pyrimidin-2-yl)acetate
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166.1800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953517 CAS-Nummer: 63155-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...