3-(Methoxymethyl)benzoic acid 95%
Lieferant: Apollo Scientific
OR965593-1G
OR965593-250MG
APOSOR965593-1GEA
106
EUR
APOSOR965593-1G
APOSOR965593-250MG
3-(Methoxymethyl)benzoic acid 95%
3-(Methoxymethyl)benzoic acid
Formel:
C₉H₁₀O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871355 CAS-Nummer: 32194-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...