4-(4-Methylphenyl)butansäure 97%

Lieferant: Apollo Scientific

Synonyme: 4-(p-Tolyl)butanoic acid

OR965734-25G
APOSOR965734-25GEA 109 EUR
APOSOR965734-25G
4-(4-Methylphenyl)butansäure 97%
4-(4-Methylphenyl)butansäure
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021786
CAS-Nummer: 4521-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR