4-(2-Pyrimidinyloxy)benzenecarbonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 4-(Pyrimidin-2-yloxy)benzonitrile

OR965738-1G OR965738-5G
APOSOR965738-1GEA 190 EUR
APOSOR965738-1G APOSOR965738-5G
4-(2-Pyrimidinyloxy)benzenecarbonitrile 97%
4-(2-Pyrimidinyloxy)benzenecarbonitrile
Formel: C₁₁H₇N₃O
Molecular Weight: 197,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01927046
CAS-Nummer: 353259-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR