4,5-Dimethylisoxazole-3-carboxylic acid 95%
Lieferant: Apollo Scientific
OR965768-1G
OR965768-250MG
APOSOR965768-1GEA
806
EUR
APOSOR965768-1G
APOSOR965768-250MG
4,5-Dimethylisoxazole-3-carboxylic acid 95%
4,5-Dimethylisoxazole-3-carboxylic acid
Formel:
C₆H₇NO₃ Molecular Weight: 141,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05667138 CAS-Nummer: 100047-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...