2-(2-Furyl)aniline 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR9658-1G
OR9658-250MG
OR9658-500MG
OR9658-5G
APOSOR9658-1GEA
393
EUR
APOSOR9658-1G
APOSOR9658-250MG
APOSOR9658-500MG
APOSOR9658-5G
2-(2-Furyl)aniline 95%
2-(2-Furyl)aniline
Formel:
C₁₀H₉NO Molecular Weight: 159,19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04039070 CAS-Nummer: 55578-79-5 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...