Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-cyclopropylthiazole-2-carboxylate
OR965854-1G
APOSOR965854-1GEA
301
EUR
APOSOR965854-1G
Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate 95%
Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate
Formel:
C₉H₁₁NO₂S Molecular Weight: 197,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11656780 CAS-Nummer: 439692-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...