3-(4-Methylpiperazin-1-yl)aniline 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(3-Aminophenyl)-4-methylpiperazine, 3-(4-Methyl-1-piperazinyl)aniline
OR9659-1G
OR9659-5G
APOSOR9659-1GEA
21.9
EUR
APOSOR9659-1G
APOSOR9659-5G
3-(4-Methylpiperazin-1-yl)aniline 98%
3-(4-Methylpiperazin-1-yl)aniline
Formel:
C₁₁H₁₇N₃ Molecular Weight: 191,28 g/mol Schmelzpunkt: 98…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08435857 CAS-Nummer: 148546-99-0 UN: 3263 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...