2-Mercapto-5-methyl-1,3,4-thiadiazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-5-thio-1,3,4-thiadiazole, 5-Methyl-1,3,4-thiadiazole-2-thiol

OR9661-1KG OR9661-250G
APOSOR9661-1KGEA 174 EUR
APOSOR9661-1KG APOSOR9661-250G
2-Mercapto-5-methyl-1,3,4-thiadiazol
2-Mercapto-5-methyl-1,3,4-thiadiazole
Formel: C₃H₄N₂S₂
Molecular Weight: 132,21 g/mol
Schmelzpunkt: 182…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038349
CAS-Nummer: 29490-19-5
EINECS: 249-667-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR