Fmoc-Cys(SO₃H)-OH·disodium salt

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-Cys(SO3H)-OH·disodium salt

OR967361-1G OR967361-5G
APOSOR967361-1GEA 259 EUR
APOSOR967361-1G APOSOR967361-5G
Fmoc-Cys(SO₃H)-OH·disodium salt
Fmoc-Cys(SO₃H)-OH·disodium salt
Formel: C₁₈H₁₅NNa₂O₇S₂
Molecular Weight: 467,418 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237016
CAS-Nummer: 163558-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR