3-Nitrobenzophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: m-Nitrobenzophenon

OR9685-25G OR9685-100G
APOSOR9685-25GEA 212 EUR
APOSOR9685-25G APOSOR9685-100G
3-Nitrobenzophenon 98%
3-Nitrobenzophenon
Formel: C₁₃H₉NO₃
Molecular Weight: 227,22 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007257
CAS-Nummer: 2243-80-3
EINECS: 218-819-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR