3-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

OR9686-1G OR9686-250MG
APOSOR9686-1GEA 249 EUR
APOSOR9686-1G APOSOR9686-250MG
3-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride 97%
3-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride
Formel: C₁₁H₈ClNOS
Molecular Weight: 237.71 g/mol
Schmelzpunkt: 52…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659063
CAS-Nummer: 844891-05-0
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR