tert-Butyl-4-(4-bromobenzyl)piperazine-1-carboxylate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(4-Bromobenzyl)piperazine, N1-BOC protected, tert-Butyl 4-(4-bromobenzyl)piperazine-1-carboxylate

OR9696-1G OR9696-250MG
APOSOR9696-1GEA 168 EUR
APOSOR9696-1G APOSOR9696-250MG
tert-Butyl-4-(4-bromobenzyl)piperazine-1-carboxylate 95%
tert-Butyl-4-(4-bromobenzyl)piperazine-1-carboxylate
Formel: C₁₆H₂₃BrN₂O₂
Molecular Weight: 355,28 g/mol
Schmelzpunkt: 69…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659068
CAS-Nummer: 844891-10-7
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR