2-Amino-5-chlorthiophenol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9714-1G
OR9714-250MG
APOSOR9714-1GEA
568
EUR
APOSOR9714-1G
APOSOR9714-250MG
2-Amino-5-chlorthiophenol 98%
2-Amino-5-chlorthiophenol
Formel:
C₆H₆ClNS Molecular Weight: 159,6 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
23474-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...