5-Nitrosalicylaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-5-nitrobenzaldehyde
OR9715-100G
OR9715-25G
OR9715-500G
APOSOR9715-100GEA
55.1
EUR
APOSOR9715-100G
APOSOR9715-25G
APOSOR9715-500G
5-Nitrosalicylaldehyd
5-Nitrosalicylaldehyd
Formel:
C₇H₅NO₄ Molecular Weight: 167,12 g/mol Schmelzpunkt: 123…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007337 CAS-Nummer: 97-51-8 EINECS: 202-587-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...