5-(3-Bromphenyl)oxazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(3-Bromophenyl)-1,3-oxazole

OR9727-5G OR9727-10G
APOSOR9727-5GEA 140 EUR
APOSOR9727-5G APOSOR9727-10G
5-(3-Bromphenyl)oxazol
5-(3-Bromophenyl)oxazole
Formel: C₉H₆BrNO
Molecular Weight: 224,06 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05668970
CAS-Nummer: 243455-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR