2-Brom-6-nitrophenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR9738-100G
OR9738-25G
OR9738-5G
APOSOR9738-100GEA
85.7
EUR
APOSOR9738-100G
APOSOR9738-25G
APOSOR9738-5G
2-Brom-6-nitrophenol 98%
2-Brom-6-nitrophenol
Formel:
C₆H₄BrNO₃ Molecular Weight: 218,01 g/mol Schmelzpunkt: 66…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683216 CAS-Nummer: 13073-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...