2-Amino-6-bromphenol 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Brom-2-hydroxyanilin
OR9740-25G
OR9740-5G
OR9740-100G
APOSOR9740-25GEA
174
EUR
APOSOR9740-25G
APOSOR9740-5G
APOSOR9740-100G
2-Amino-6-bromphenol 97+%
2-Amino-6-bromphenol
Formel:
C₆H₆BrNO Molecular Weight: 188,02 g/mol Schmelzpunkt: 83…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08277300 CAS-Nummer: 28165-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...