3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonyl chloride 97%

Lieferant: Apollo Scientific

OR9756-10G OR9756-1G OR9756-5G
APOSOR9756-10GEA 1040 EUR
APOSOR9756-10G APOSOR9756-1G APOSOR9756-5G
3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonyl chloride 97%
3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonyl chloride
Formel: C₁₂H₁₁ClN₂O
Schmelzpunkt: 57,5…58.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03421620
CAS-Nummer: 61226-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR