1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester
OR9760-250MG
OR9760-1G
APOSOR9760-250MGEA
21.9
EUR
APOSOR9760-250MG
APOSOR9760-1G
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine 95%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine
Formel:
C₁₆H₂₄BNO₂ Molecular Weight: 273.18 g/mol Siedepunkt: 396 °C Schmelzpunkt: 108.5…109.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08060504 CAS-Nummer: 852227-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...