4-Amino-3-chlorbenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR9762-100G
OR9762-25G
OR9762-5G
APOSOR9762-100GEA
133
EUR
APOSOR9762-100G
APOSOR9762-25G
APOSOR9762-5G
4-Amino-3-chlorbenzonitril
4-Amino-3-chlorbenzonitril
Formel:
C₇H₅ClN₂ Molecular Weight: 152,58 g/mol Schmelzpunkt: 100…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052917 CAS-Nummer: 21803-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...