3-Amino-2-chlor-4-methylpyridin

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Amino-2-chloro-4-methylpyridine, 3-Amino-2-chloro-4-picoline, 2-Chloro-4-methylpyridin-3-amine

OR9771-100G OR9771-25G OR9771-5G
APOSOR9771-100GEA 26.1 EUR
APOSOR9771-100G APOSOR9771-25G APOSOR9771-5G
3-Amino-2-chlor-4-methylpyridin
3-Amino-2-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142.59 g/mol
Schmelzpunkt: 64…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00673152
CAS-Nummer: 133627-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR