1-(4-Nitrophenyl)-1,2-propandione 95+%

Lieferant: Apollo Scientific
Achtung

OR9779-1G OR9779-250MG
APOSOR9779-1GEA 56.7 EUR
APOSOR9779-1G APOSOR9779-250MG
1-(4-Nitrophenyl)-1,2-propandione 95+%
1-(4-Nitrophenyl)-1,2-propandione
Formel: C₉H₇NO₄
Molecular Weight: 193,16 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01882876
CAS-Nummer: 6159-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR