Ethyl-2-[(aminocarbonyl)amino]acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl ureidoacetate

OR9785-1G OR9785-5G
APOSOR9785-1GEA 139 EUR
APOSOR9785-1G APOSOR9785-5G
Ethyl-2-[(aminocarbonyl)amino]acetate
Ethyl-2-[(aminocarbonyl)amino]acetate
Formel: C₅H₁₀N₂O₃
Molecular Weight: 146,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025456
CAS-Nummer: 6293-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR