3-Butoxybenzoic acid

Lieferant: Apollo Scientific

OR9793-1G OR9793-500MG OR9793-5G
APOSOR9793-1GEA 140 EUR
APOSOR9793-1G APOSOR9793-500MG APOSOR9793-5G
3-Butoxybenzoic acid
3-Butoxybenzoic acid
Formel: C₁₁H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01143605
CAS-Nummer: 93351-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR