4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-((1H-1,2,4-Triazol-1-yl)methyl)benzonitrile
OR9802-1G
OR9802-5G
APOSOR9802-1GEA
58.4
EUR
APOSOR9802-1G
APOSOR9802-5G
4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile 97%
4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile
Formel:
C₁₀H₈N₄ Molecular Weight: 184.2 g/mol Schmelzpunkt: 69…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368016 CAS-Nummer: 112809-25-3 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...