Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-amino-5-bromo-3-methylbenzoate
OR9818-1G
OR9818-5G
APOSOR9818-1GEA
59.6
EUR
APOSOR9818-1G
APOSOR9818-5G
Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate
Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244,09 g/mol Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797379 CAS-Nummer: 206548-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...