4-Hydroxy-2-phenyl-5-pyrimidinecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR9823-1G
OR9823-250MG
APOSOR9823-1GEA
212
EUR
APOSOR9823-1G
APOSOR9823-250MG
4-Hydroxy-2-phenyl-5-pyrimidinecarboxylic acid
4-Hydroxy-2-phenyl-5-pyrimidinecarboxylic acid
Formel:
C₁₁H₈N₂O₃ Schmelzpunkt: 269…272 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01411768 CAS-Nummer: 56406-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...