2-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine 95+%

Lieferant: Apollo Scientific

Synonyme: 2-(tert-Butyl)-7-azaindole

OR9836-10G OR9836-1G OR9836-5G
APOSOR9836-10GEA 575 EUR
APOSOR9836-10G APOSOR9836-1G APOSOR9836-5G
2-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine 95+%
2-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₁₁H₁₄N₂
Molecular Weight: 174.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02083381
CAS-Nummer: 86847-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR