2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one 95%

Lieferant: Apollo Scientific
Achtung

OR9861-1G OR9861-5G OR9861-10G
APOSOR9861-1GEA 21.9 EUR
APOSOR9861-1G APOSOR9861-5G APOSOR9861-10G
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one 95%
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one
Formel: C₇H₆N₂O₂
Molecular Weight: 150,14 g/mol
Schmelzpunkt: 204…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006697
CAS-Nummer: 20348-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR