2,3-Diphenyl-6-nitrochinoxalin

Lieferant: Apollo Scientific
Gefahr

OR9865-1G OR9865-2.5G
APOSOR9865-1GEA 148 EUR
APOSOR9865-1G APOSOR9865-2.5G
2,3-Diphenyl-6-nitrochinoxalin
2,3-Diphenyl-6-nitroquinoxaline
Formel: C₂₀H₁₃N₃O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 7466-45-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR