4-Piperidin-1-ylbenzylamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: [4-(Piperidin-1-yl)phenyl]methylamine

OR9881-1G OR9881-250MG
APOSOR9881-1GEA 396 EUR
APOSOR9881-1G APOSOR9881-250MG
4-Piperidin-1-ylbenzylamine
4-Piperidin-1-ylbenzylamine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659080
CAS-Nummer: 214759-73-6
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR