Methyl-2-(3-amino-4-hydroxyphenyl)acetate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-(3-amino-4-hydroxyphenyl)acetate

OR9882-1G OR9882-250MG OR9882-5G
APOSOR9882-1GEA 69.6 EUR
APOSOR9882-1G APOSOR9882-250MG APOSOR9882-5G
Methyl-2-(3-amino-4-hydroxyphenyl)acetate 95%
Methyl-2-(3-amino-4-hydroxyphenyl)acetate
Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
Schmelzpunkt: 102…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04125316
CAS-Nummer: 78587-72-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR