Imidazo[1,2-a]pyridin-8-ol 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Hydroxyimidazo[1,2-a]pyridine

OR9887-500MG
APOSOR9887-500MGEA 143 EUR
APOSOR9887-500MG
Imidazo[1,2-a]pyridin-8-ol 95+%
Imidazo[1,2-a]pyridin-8-ol
Formel: C₇H₆N₂O
Molecular Weight: 134,1400 g/mol
Schmelzpunkt: 154…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06496222
CAS-Nummer: 69214-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR