ω,4-Dibromacetophenon 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Brom-1-(4-bromphenyl)-1-ethanon, Tau Protein Kinase I Inhibitor, 2,4'-Dibromacetophenon, Glycogen Synthase Kinase-3β Inhibitor VII, GSK-3β Inhibitor VII, 4-Bromphenacylbromid, TPK I Inhibitor, α,4-Dibromacetophenon
OR9901-100G
OR9901-25G
APOSOR9901-100GEA
133
EUR
APOSOR9901-100G
APOSOR9901-25G
ω,4-Dibromacetophenon 98%
ω,4-Dibromacetophenon
Formel:
C₈H₆Br₂O Molecular Weight: 277,94 g/mol Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000200 CAS-Nummer: 99-73-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...