ω-Brom-4-iodacetophenon 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-1-(4-iodophenyl)ethanone, 4-Iodophenacyl bromide
OR9902-1G
OR9902-25G
OR9902-5G
APOSOR9902-1GEA
21.9
EUR
APOSOR9902-1G
APOSOR9902-25G
APOSOR9902-5G
ω-Brom-4-iodacetophenon 97%
ω-Brom-4-iodacetophenon
Formel:
C₈H₆BrIO Molecular Weight: 324,9400 g/mol Schmelzpunkt: 113…115 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01360521 CAS-Nummer: 31827-94-8 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...