2-Brom-4'-(methylthio)acetophenon
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Methylthio)phenacyl bromide, 2-Bromo-1-(4-(methylthio)phenyl)ethanone
OR9903-1G
OR9903-5G
APOSOR9903-1GEA
65.4
EUR
APOSOR9903-1G
APOSOR9903-5G
2-Brom-4'-(methylthio)acetophenon
2-Brom-4'-(methylthio)acetophenon
Formel:
C₉H₉BrOS Molecular Weight: 245.136 g/mol Schmelzpunkt: 50…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094708 CAS-Nummer: 42445-46-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...