3-Amino-6-brom-1H-indazol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-1H-indazol-3-amine

OR9913-1G OR9913-5G OR9913-25G
APOSOR9913-1GEA 21.9 EUR
APOSOR9913-1G APOSOR9913-5G APOSOR9913-25G
3-Amino-6-brom-1H-indazol 95%
3-Amino-6-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 237…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05665872
CAS-Nummer: 404827-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR