3-Amino-5-brom-1H-indazol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1H-indazol-3-amine

OR9914-1G OR9914-5G OR9914-25G
APOSOR9914-1GEA 21.9 EUR
APOSOR9914-1G APOSOR9914-5G APOSOR9914-25G
3-Amino-5-brom-1H-indazol 95%
3-Amino-5-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 171…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426696
CAS-Nummer: 61272-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR