Cumarin-3-carbonsäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Oxo-2H-chromene-3-carboxylic acid
OR9923-25G
OR9923-5G
OR9923-100G
OR9923-500G
APOSOR9923-25GEA
72.5
EUR
APOSOR9923-25G
APOSOR9923-5G
APOSOR9923-100G
APOSOR9923-500G
Cumarin-3-carbonsäure 97%
Cumarin-3-carbonsäure
Formel:
C₁₀H₆O₄ Molecular Weight: 190,16 g/mol Schmelzpunkt: 189…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006852 CAS-Nummer: 531-81-7 EINECS: 208-518-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...