1-(4-Bromphenyl)-2-pyrrolidinon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromophenyl)pyrrolidin-2-one

OR9934-1G OR9934-5G OR9934-250MG
APOSOR9934-1GEA 72.5 EUR
APOSOR9934-1G APOSOR9934-5G APOSOR9934-250MG
1-(4-Bromphenyl)-2-pyrrolidinon
1-(4-Bromphenyl)-2-pyrrolidinon
Formel: C₁₀H₁₀BrNO
Molecular Weight: 240,10 g/mol
Schmelzpunkt: 98…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00138502
CAS-Nummer: 7661-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR