3-Chlor-6-hydroxy-2,4-dimethylbenzaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-2,4-dimethyl-6-hydroxybenzaldehyde
OR9955-250MG
OR9955-100MG
APOSOR9955-250MGEA
98.6
EUR
APOSOR9955-250MG
APOSOR9955-100MG
3-Chlor-6-hydroxy-2,4-dimethylbenzaldehyd 95%
3-Chlor-6-hydroxy-2,4-dimethylbenzaldehyd
Formel:
C₉H₉ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00732125 CAS-Nummer: 81322-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...