2-Chlor-5-nitrophenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9957-100G
APOSOR9957-100GEA
119
EUR
APOSOR9957-100G
2-Chlor-5-nitrophenol 97%
2-Chlor-5-nitrophenol
Formel:
C₆H₄ClNO₃ Molecular Weight: 173,56 g/mol Schmelzpunkt: 112…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01571825 CAS-Nummer: 619-10-3 EINECS: 210-579-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...