(-)-Cytisin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(1R,5S)-1,2,3,4,5,6-Hexahydro-8H-1,5-methanopyrido[1,2-a][1,5]diazocin-8-one, (1R,5S)-3,4,5,6-Tetrahydro-1H-1,5-methanopyrido[1,2-a][1,5]diazocin-8(2H)-one, Cytisine, Cytisinicline
OR9985T-5G
APOSOR9985T-5GEA
115
EUR
APOSOR9985T-5G
(-)-Cytisin
(-)-Cytisine
Toxic principle in seed of Laburnum anagyroides & other Leguminosae.
Formel:
C₁₁H₁₄N₂O Molecular Weight: 190,25 g/mol Siedepunkt: 218 °C Schmelzpunkt: 154…1576 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136048 CAS-Nummer: 485-35-8 EINECS: 207-616-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...