(2-Chlor-3,6-difluorphenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

PC0017-1G PC0017-5G PC0017-100MG
APOSPC0017-1GEA 116 EUR
APOSPC0017-1G APOSPC0017-5G APOSPC0017-100MG
(2-Chlor-3,6-difluorphenyl)essigsäure
(2-Chlor-3,6-difluorphenyl)essigsäure
Formel: C₈H₅ClF₂O₂
Molecular Weight: 206.58 g/mol
Schmelzpunkt: 136…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631448
CAS-Nummer: 261762-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR